Inlet base seals
Supplier: Thermo Fisher Scientific
290GA083 290GA084 290GA091 290GA092 290GA094 290GA122 290GA082 290GA081
555-9130EA 109 EUR
555-9130 555-9223 555-9222 555-9221 555-9219 555-9215 555-9037 555-9038
Inlet base seals
Seals Mechanical Seals
Ideal for analysis of active compounds such as pesticides, polychlorinated biphenyls (PCBs) and phenol.
Compatible with all Agilent GC 5890/6890/7890 split/splitless injection ports.
Order Now
SPECIFICATIONS
Environmentally Preferable