Non-return valve
Supplier: Lenz Laborglas GmbH & CO.KG
05590000
5590000.EA 35.7 EUR
5590000.
Non-return valve
Valves Non Return Valves
DURAN®, borosilicate glass, clear. Prevent pressure loss or backflow. Simple design, cannot be dismantled.
  • Excellent chemical resistance
  • Excellent thermal resistance, autoclavable
  • With tapered ends to ensure that the tube is held firmly