J.T.Baker® BAKER ANALYZED®, Buffer Solutions
Supplier: Avantor
7261.1000EA 58.9 EUR
7261.1000 7262.1000 7267.1000 7263.1000 7265.1000 7268.1000 7270.1000 5678.1000 5677.1000 5643.1000 5609.0500 5609.1000 5608.9010 5608.5000PE 5608.2500PE 5608.1000 5608.0500 5606.9010 5606.5000PE 5606.2500PE 5606.1000 5606.0500 5609.9010
J.T.Baker® BAKER ANALYZED®, Buffer Solutions
Buffers pH Calibration Buffers
Ready-to-use pH buffers are manufactured in large lots to stringent specifications and utilise quality control procedures to reduce lot-to-lot variability. You can eliminate the time and expense of preparation and standardisation of solutions, helping streamline high-volume analytical work in applications such as quality control.
Contain 0,01% Sodium benzoate.
Certifications: This buffer is traceable to NIST Standard reference buffer.
Order Now
SPECIFICATIONS
Environmentally Preferable