Hydrochloric acid 0.1 mol/l (0.1 N), BAKER ANALYZED®, Laboratory Reagent volumetric solution, J.T.Baker®

Supplier: Avantor
Warning

Synonyms: Hydrochloric acid solution, Muriatic acid

Management of Change (MOC) category = NI

5621-02EA 70.55 EUR
5621-02
Hydrochloric acid 0.1 mol/l (0.1 N), BAKER ANALYZED®, Laboratory Reagent volumetric solution, J.T.Baker®
Hydrochloric acid

J.T.Baker® volumetric and analytical solutions. These solutions are suitable for use in ACS, USP and NF compendial methods and general laboratory applications. 

Formula: HCl
MW: 36.46 g/mol
Density: 1.004 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011324
CAS Number: 7647-01-0
EINECS: 231-595-7
UN: 1789
ADR: 8 III

Order Now

Specification Test Results

For Laboratory,Research,or Manufacturing Use 0.0995 - 0.1005
Normality 0.0995 - 0.1005
Sulfate (SO₄) ≤ 1 ppm
Ammonium (NH₄) ≤ 3 ppm
Trace Impurities - Heavy Metals ≤ 0.5 ppm
Trace Impurities - Iron (Fe) ≤ 0.1 ppm
Standardization at 25°C traceable to NIST Standard Reference Material.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR