L(-)-Sorbose, Millipore®

Supplier: Merck Millipore Calbiochem

Synonyms: L-(-)-Sorbose, (3S,4R,5S)-1,3,4,5,6-Pentahydroxyhexan-2-one

567692-1 567692-250
567692-1EA 2640 EUR
567692-1 567692-250
L(-)-Sorbose, Millipore®
L(-)-Sorbose
Formula: C₆H₁₂O₆
MW: 180.16 g/mol
Boiling Pt: 380
Melting Pt: 158…160 °C
Storage Temperature: Ambient
MDL Number: MFCD00151097
CAS Number: 87-79-6
EINECS: 201-771-8
Merck Index: 12,08874

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR