Furnace tong, Ward pattern
Supplier: R L ENTERPRISES
5704
5704.EA 504 EUR
5704.
Furnace tong, Ward pattern
Tongs
This tong is used for protection and safety when handling heated objects in the laboratory.
  • Manufactured from stainless steel
  • For handling a wide variety of crucibles, especially in muffle furnaces
  • Operated by thumb action with one hand
Fitted with a circular shield to protect the hand against heat from the furnace.