J.T.Baker®, Bismuth Standard Solution, 10000 mg/L Bi in 10% Nitric Acid (from Bi) Standard for ICP
Supplier: Avantor
5721.0100EA 0 EUR
5721.0100
J.T.Baker®, Bismuth Standard Solution, 10000 mg/L Bi in 10% Nitric Acid (from Bi) Standard for ICP
Standards Cationic Standards Bismuth Standards
Certifications: Traceable to NIST