J.T.Baker®, Boron Standard Solution, 10000 mg/L B in Water 2% Ammonium Hydroxide (from B(OH₃)) Standard for ICP
Supplier: Avantor
5722.0100EA 0 EUR
5722.0100
J.T.Baker®, Boron Standard Solution, 10000 mg/L B in Water 2% Ammonium Hydroxide (from B(OH₃)) Standard for ICP
Standards Cationic Standards Boron Standards
Certifications: Traceable to NIST