Adaptors for exhaust filter connection, b.safe
Supplier: Bohlender
M516-01 M516-05
590-0202EA 33.4 EUR
590-0202 590-0203
Adaptors for exhaust filter connection, b.safe
Fittings and Connectors
Adaptor for installation of capillaries with b.safe fittings at the exhaust filter connection of the b.safe waste caps.
  • Excellent chemical resistance
  • Converts the exhaust filter port into a capillary port
  • Suitable for different capillary diameters
Suitable b.safe fittings for different capillary diameters are separately available.