HPLC columns, SUPELCOSIL™, Suplex™ pKb-100, Supelco®

Supplier: Merck
58934 58932 59531-U 59541-U 59172
58934.EA 692 EUR
58934. 58932. 59531-U 59541-U 59172.
HPLC columns, SUPELCOSIL™, Suplex™ pKb-100, Supelco®
Chromatography Columns
Suplex pKb-100 columns feature the same bonded phase functionality as SUPELCOSIL LC-ABZ columns. These specially deactivated columns differ in that Suplex pKb-100 is not endcapped, while SUPELCOSIL LC-ABZ is endcapped. The absence of the end-capping reagent results in better performance from Suplex pKb-100 for the strongest basic compounds, while LC-ABZ is preferred when the sample also contains acids and/or zwitterions.

  • Alkylamide phase
  • Silica gel particle platform

Surface area: 170 m²/g

Carbon load: 12,5%

pH range: 2 to 7,5
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR