Magnesium chloride hexahydrate 99.0-102.0%, crystals, AR® ACS, Macron Fine Chemicals™

Supplier: Avantor Performance Materials

5958-04EA 119 EUR
5958-04 5958-26
Magnesium chloride hexahydrate 99.0-102.0%, crystals, AR® ACS, Macron Fine Chemicals™
Magnesium chloride hexahydrate

ACS—Reagents meeting the requirements of the American Chemical Society Committee on Analytical Reagents. The specifications listed in this catalog are based on the ACS Reagent Chemicals Tenth Edition.  

Formula: MgCl₂·6H₂O
Boiling Pt: 1412 °C (1013 hPa)
Melting Pt: 117 °C
Density: 1,57 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00149781
CAS Number: 7791-18-6
EINECS: 232-094-6
Merck Index: 13,05684

Order Now

Specification Test Results

Meets ACS Reagent Chemical Requirements
For Laboratory,Research,or Manufacturing Use 99.0 - 102.0%
ACS - Assay (MgCl₂·6H₂O) 99.0 - 102.0 %
ACS - Ammonium (NH₄) ≤0.001 %
ACS - Barium (Ba) ≤0.005 %
ACS - Calcium (Ca) ≤0.01 %
ACS - Heavy Metals (as Pb) ≤5 ppm
ACS - Insoluble Matter ≤0.005 %
ACS - Iron (Fe) ≤5 ppm
ACS - Manganese (Mn) ≤5.000 ppm
ACS - Nitrate (NO₃) ≤0.001 %
ACS - pH of 5% Solution 4.5 - 7.0
ACS - Phosphate (PO₄) ≤5 ppm
ACS - Potassium (K) ≤0.005 %
ACS - Sodium (Na) ≤0.005 %
ACS - Strontium (Sr) ≤0.005 %
ACS - Sulfate (SO₄) ≤0.002 %
The net weight of the product in each container is within +/- 2% of the netweight stated on the label.

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR