Ammonia solution 28%, BAKER ANALYZED® ACS, J.T.Baker®

Supplier: Avantor
Danger

6005.1000EA 44.41 EUR
6005.1000 6005.2500 6005.9025
Ammonia solution 28%, BAKER ANALYZED® ACS, J.T.Baker®
Ammonia solution
Formula: NH₃
Boiling Pt: 32…37,7 °C (1013 hPa)
Melting Pt: –60…–57,5 °C
Density: 0,88…0.91 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00066650
CAS Number: 1336-21-6
EINECS: 215-647-6
UN: 2672
ADR: 8,III

Order Now

Specification Test Results

Exceeds ACS Specifications
Assay (as NH3) (acidimetric) 28.0 - 30.0 %
Appearance passes test
Carbon Dioxide (CO2) max. 0.002 %
Color (APHA) max. 5
Chloride (Cl) max. 0.5 ppm
Residue after Ignition max. 0.001 %
Specific Gravity at 60°/60°F 0.896 - 0.902
Substances Reducing KMnO4 passes test
Trace Impurities (in ppm)
Arsenic and Antimony (as As) max. 3
Aluminium (Al) max. 0.2
Boron (B) max. 0.05
Barium (Ba) max. 0.3
Chromium (Cr) max. 0.1
Copper (Cu) max. 0.1
Iron (Fe) max. 0.1
Magnesium (Mg) max. 0.2
Manganese (Mn) max. 0.1
Nickel (Ni) max. 0.1
Lead (Pb) max. 0.2
Tin (Sn) max. 0.1
Titanium (Ti) max. 0.1
Zinc (Zn) max. 0.1
Heavy Metals (as Pb) max. 0.5
Nitrate (NO3) max. 2
Phosphate (PO4) max. 0.4
Sulfate (SO4) max. 2


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR