Potassium carbonate, anhydrous ≥99.0% (by titrimetric analysis), Puriss. p.a. ACS, Fluka™

Supplier: Honeywell Chemicals
Warning

60109-4X2.5KG 60109-2.5KG 60109-250G 60109-1KG 60109-25KG 60109-500G
60109-4X2.5KGEA 610 EUR
60109-4X2.5KG 60109-2.5KG 60109-250G 60109-1KG 60109-25KG 60109-500G
Potassium carbonate, anhydrous ≥99.0% (by titrimetric analysis), Puriss. p.a. ACS, Fluka™
Potassium carbonate
Formula: K₂CO₃
Boiling Pt: Decomposition
Melting Pt: 891 °C
Density: 2,430 g/cm³ (19 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011382
CAS Number: 584-08-7
EINECS: 209-529-3
Merck Index: 13,07702
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4d5900faa1ba52c3213eed8f6bb2d952bac1f30f&vid=6298361525e6af416556b5694c74a8a91faa437c&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay 99.0 - 101.0 %
Loss on drying (300°C) Max. 1 %
Water insoluble matter Max. 0.01 %
Silver (Ag) Max. 5 ppm
Aluminium (Al) Max. 5 ppm
Arsenic (As) Max. 0.5 ppm
Barium (Ba) Max. 5 ppm
Bismuth (Bi) Max. 5 ppm
Calcium (Ca) Max. 10 ppm
Cadmium (Cd) Max. 5 ppm
Cobalt (Co) Max. 5 ppm
Chromium (Cr) Max. 5 ppm
Copper (Cu) Max. 5 ppm
Iron (Fe) Max. 5 ppm
Lithium (Li) Max. 5 ppm
Magnesium (Mg) Max. 5 ppm
Manganese (Mn) Max. 5 ppm
Molybdenum (Mo) Max. 5 ppm
Sodium (Na) Max. 200 ppm
Nickel (Ni) Max. 5 ppm
Lead (Pb) Max. 5 ppm
Strontium (Sr) Max. 5 ppm
Thallium (Tl) Max. 5 ppm
Zinc (Zn) Max. 5 ppm
Total S (as SO₄) Max. 40 ppm
Heavy metals (as Pb) Max. 5 ppm
Silicate (as SiO₂) Max. 50 ppm
Chloride (Cl) Max. 30 ppm
Phosphate (PO₄) Max. 10 ppm
Total nitrogen (as N) Max. 10 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR