Nitric acid 70% 70, BAKER ANALYZED® ACS, J.T.Baker®

Supplier: Avantor
Danger

Synonyms: Nitric acid solution

6020.2500PEEA 145.02 EUR
6020.2500PE
Nitric acid 70% 70, BAKER ANALYZED® ACS, J.T.Baker®
Nitric acid
Formula: HNO₃
MW: 63.01 g/mol
Boiling Pt: 87
Melting Pt: -42
Density: 1.51
Storage Temperature: Ambient
MDL Number: MFCD00011349
CAS Number: 7697-37-2
EINECS: 231-714-2
UN: 2031
ADR: 8 I
Merck Index: 13,06610

Order Now

Specification Test Results

Exceeds ACS Specifications. Meets Reagent Specifications for testing USP/NF monographs.
Assay (acidimetric) 69.0 - 70.0 %
Appearance passes test
Color (APHA) max. 10
Residue after Ignition max. 4 ppm
Specific Gravity at 60°/60°F 1.416 - 1.420
Trace Impurities (in ppm)
Trace Impurities (in ppm) Chloride (Cl) max. 0.1
Trace Impurities (in ppm)Phosphate (PO4) max. 0.2
Trace Impurities (in ppm)Sulfate (SO4) max. 0.5
Trace Impurities (in ppb)
Arsenic and Antimony (as As) max. 4
Aluminium (Al) max. 100
Gold (Au) max. 200
Boron (B) max. 50
Calcium (Ca) max. 200
Chromium (Cr) max. 100
Copper (Cu) max. 50
Iron (Fe) max. 100
Mercury (Hg) max. 5
Potassium (K) max. 300
Magnesium (Mg) max. 100
Manganese (Mn) max. 100
Nickel (Ni) max. 50
Lead (Pb) max. 100
Tin (Sn) max. 200
Titanium (Ti) max. 200
Zinc (Zn) max. 200
Heavy Metals (as Pb) max. 100


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR