J.T.Baker® BAKERBOND® PolyQUAT™, Multimode Ion Exchange (IEX) Chromatography Resin, Lab Column
Supplier: Avantor
6065-07EA 331.98 EUR
6065-07 6065-25
J.T.Baker® BAKERBOND® PolyQUAT™, Multimode Ion Exchange (IEX) Chromatography Resin, Lab Column
Chromatography Columns
The J.T.Baker® BAKERBOND® PolyQUAT™ multimode ion exchange (IEX) chromatography resin functions as a strong anion exchange resin with improved selectivity for superior separation in process chromatography applications.
Powered by a proprietary surface chemistry combining strong anion exchange functionality with weak anionic exchange sites and hydrophilicity, BAKERBOND® PolyQUAT™ multimode cation exchange chromatography media utilizes its unique selectivity to enable higher purification performance across a wide range of biopharmaceuticals.
Designed and manufactured by Avantor to theMore Product Information