Silica gel, Pore size: 100 Å 15-25 μm, high_purity Chromatography_column, Supelco®

Supplier: Merck


MQ100

60744-500G
60744-500GEA 261 EUR
60744-500G
Silica gel, Pore size: 100 Å 15-25 μm, high_purity Chromatography_column, Supelco®
Silica gel
Formula: SiO₂
MW: 60.08 g/mol
Boiling Pt: 2230
Melting Pt: 1710
Storage Temperature: Ambient
MDL Number: MFCD00011232
CAS Number: 112926-00-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b52a66798cf52d4e9a43a6bb4cf37d745498fd9f&vid=45ad1e309e32ee57f381073e8047905577669924&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR