VWR®, Semi-Micro Balances with Ioniser, SM and SMG Series
Supplier: VWR Collection
611-3801EA 2680 EUR
611-3801 611-3803 611-3804 611-3806 611-3807 611-3809 611-3810 611-3800 611-4284 611-4294 611-4314 611-4315
VWR®, Semi-Micro Balances with Ioniser, SM and SMG Series
Balances and Scales
Semi-micro balances with resolution of 0,01 mg and a built-in ioniser, which ensures accurate sample preparation. Static charges accumulate in different samples that are routinely weighed in the laboratory. These static charges can build up and alter the weighing value. By blowing ions, the built-in ioniser neutralises static electricity and within seconds the sample is ready to be weighed. The balance calibration system automatically calibrates the balance when there is a temperature change or at a set period of time (factory settings), this ensures accurate measurements even if the userMore Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable