Disposable weighing dishes
Supplier: LUDI
176154710
611-9000EA 37.3 EUR
611-9000
Disposable weighing dishes
Boats Weighing Boats
Aluminium.
  • Diameter: 100 mm
  • Height of rim: 7 mm
For moisture determination.