Disposable weighing boats, VWR®
Supplier: VWR Collection
611-0097EA 60.5 EUR
611-0097 611-9184 611-9185 611-9186 611-9187 611-9188 611-9189 611-9190 611-9191 611-9192 611-9161 611-9162 611-1993 611-1994 611-1995 611-1996
Disposable weighing boats, VWR®
Boats Weighing Boats
PS, diamond.
  • Wide, flat bottom avoids tipping
  • Temperature resistant up to +70 °C
These flexible boats feature smooth surfaces to provide accurate pour-out with minimal sample loss. They are ideal for use with microwaves and moisture analysers.
Packaging: They are produced and packed in plastic bags in a cleanroom.
Order Now
SPECIFICATIONS
Environmentally Preferable