Pasteur pipettes, VOLAC®
Supplier: Poulten & Graf
D810 D812 D810/PP D812/PP
D810..EA 197 EUR
D810.. D812.. 612-0994 612-1103
Pasteur pipettes, VOLAC®
Pipettes Pasteur/Transfer Pipettes
Glass, graduated, plain or pre-plugged, sterile or non sterile.
  • Precision pipettes with a heavy wall thickness to provide the best quality product allied with greater strength to give lower breakages during transit, and whilst in use or, when fitting PVC teats
  • Standard pipettes with top body, outer diameter 6,8 to 7,1 mm, wall thickness 0,50 to 0,53 mm and jet outer diameter 1,50 ±0,2 mm
Order Now
SPECIFICATIONS
Environmentally Preferable