Serological pipettes
Supplier: Greiner Bio-One
604181_1000 606180_200 607180_200 710180_1000 760180_200 607107_500 710107_1000 604107_1000 768180_100 606107_500 768160_100 607160_200 760160_200 606160_200 604160_1000 710160_1000 760107_200 760160-TRI_100
612-1011EA 411 EUR
612-1011 612-1013 612-1015 612-1012 612-1017 612-1020 612-9107 612-0677 612-1018 612-1019 612-3328 612-3325 612-3327 612-3324 612-3272 612-3323 GREI760107_200 GREI760160-TRI_100
Serological pipettes
Pipettes Serological Pipettes
High-quality transparent PS, graduated, sterile.
  • Clear and easy-to-read graduation, with a vertical coloured stripe on the 1, 2, 5, 10 and 25-ml pipettes
  • Increased capacity thanks to negative graduation
  • Cotton insert to stop liquids from rising up into the pipetting device
  • Colour code conforms to international standards to facilitate pipette selection
  • Free from DNase, RNase and human DNA, non pyrogenic and non cytotoxic
Packaging: Bulk packed in plastic resealable zip sachets, individually wrapped in peelable plastic/paper, or individually packaged inMore Product Information