Graduated pipettes, SILBERBRAND ETERNA, type 3, Class B, total delivery
Supplier: Brand
27070 27072 27073 27075 27076 27077 27078 27079
612-1145EA 7.7 EUR
612-1145 612-1148 612-1149 612-1152 612-1153 612-1154 612-1155 612-1156
Graduated pipettes, SILBERBRAND ETERNA, type 3, Class B, total delivery
Pipettes Graduated Pipettes
AR-Glas®.
  • Calibrated to deliver (TD, Ex)
  • Marks and inscriptions in ETERNA amber stain
  • Marked pipettes (*) suitable for cotton plugs
DIN EN ISO 835
Length: 360 ±10 mm except for 25 ml pipettes: 450 ±10 mm.
Certifications: DIN EN ISO 835.