Burette self-zero kit, Nalgene®
Supplier: Thermo Fisher Scientific
3645-2550
612-2587EA 67.9 EUR
612-2587
Burette self-zero kit, Nalgene®
Burettes
Makes any plastic or glass burette self-zeroing.
Consists of a 1000 ml LDPE bottle, 1,2 m clear PVC tubing, PP fill tube and adapter.
Order Now
SPECIFICATIONS
Environmentally Preferable Low Manufacturing Impact
Environmental certification NAR Certificate of Retirement