Pipette tips, Finntip™
Supplier: Thermo Fisher Scientific
9400220 9400230 9400250 9400260 9400263 9400300 9400303 9400310 9400370 9400360 9400610 9400613 9400620 9401030 9401250 9401253 9401255 9401260 9402030 9402050 9402151 9402163 9400267 9401115 9401110 9401113 9402073 9402070 9400327 9400373 9402171 9402180 9402183
613-2594EA 0 EUR
613-2594 613-2595 613-2596 613-2597 613-2598 613-2599 613-2600 613-2601 612-6346 613-2603 613-2604 613-2605 613-2606 613-2608 613-2612 613-2613 613-2614 613-2615 613-2622 613-2623 613-2627 613-2629 612-6343 613-4485 612-6378 613-4484 613-5010 613-5009 612-2508 612-6353 613-7437 613-7438 613-7439
Pipette tips, Finntip™
Pipette Tips
Finntip™ pipette tips are designed to achieve the highest level of performance in terms of accuracy and repeatability in conjunction with Finnpipette™ pipettes.
  • High quality PP, autoclavable
  • Unique shape of the collar ensures a leak-free connection to the pipette
  • Smooth, hydrophobic surface minimises liquid retention allowing total dispensing of the sample
  • Colour coded racks for rapid identification of the tips to be used
  • Sterile tips: Certified RNase-, DNase-, DNA- and endotoxin-free
Packaging: Available in different volumes and packed loose, inMore Product Information