VWR®, Pipette Tips
Supplier: VWR Collection
613-0252EA 0 EUR
613-0252 613-0277 613-0260 613-0265 613-0296 613-0304 732-0505 613-0335 613-0342 613-0368 613-0369 613-0637 613-0638 613-0831 613-0892 613-1634
VWR®, Pipette Tips
Pipette Tips
These tips are made of high quality, 100% pure, virgin medical grade PP. Made in the highest quality moulds, these tips can be used whenever the work demands a higher standard.
  • Sustainable design based on virgin resins
  • A wide seal ensures tips will seal easily and consistently without leaking
  • Guaranteed free of detectable DNA, DNase/RNase, endotoxins and heavy metals
  • Tested for endotoxin (pyrogen) contamination
  • Resins are pre-tested for metal contamination
  • Graduated tips only: Graduations allow a visual check of pipetting accuracy
  • Certificates are on file
More Product Information