Filter tips, Finntip™ Flex
Supplier: Thermo Fisher Scientific
94056510 94056520 94056580 94056710 94056810 94056980 94056970 94056380
613-0821EA 151 EUR
613-0821 613-0822 613-0823 613-0824 613-0825 613-0820 613-0804 613-0802
Filter tips, Finntip™ Flex
Pipette Tips
Sterile PP tips with PE filter.
  • Free from DNA, DNase and RNase
  • Filters are made of an inert non sealing polyethylene matrix
  • Filters are effective at eliminating carryover contamination from liquids and aerosols between pipetter and tip
  • Supplied in vacuum-sealed sterilised tip racks
Finntip™ Flex filter tips combine the ergonomics of a standard Finntip Flex pipette tip and the safety of a filter tip. Finntip Flex tips are especially designed to enhance pipetting ergonomics. With its thin and flexible tip walls and special rack design, the tipMore Product Information