VWR®, Dispenser Tips, Classic
Supplier: VWR Collection
613-1000EA 111 EUR
613-1000 613-1001 613-1002 613-1003 613-1004 613-1005 613-1006 613-1007 613-1008 613-1009 613-1011 613-1012 613-1013
VWR®, Dispenser Tips, Classic
Pipette Tips
By using the direct displacement method, these tips work well for infectious and volatile liquids, as the fluid never will get in touch with the instrument which avoids any contamination.
  • Compatible with: VWR Stepper and Stepper Pro, Ritter® Ripette® and Ripette® pro, Brand® HandyStep® and HandyStep® electronic*, Eppendorf® Multipette® 4780, Gilson Repetman® and Easystep (* without automatic tip recognition)
  • Sustainable design based on virgin resins
  • Eight different tip sizes from 0,05 to 50 ml
  • Sterile tips guaranteed free of DNA, DNase, RNase,
More Product Information