Robotic tips for BioTek™ pipetting systems
Supplier: Thermo Fisher Scientific
986-021
613-1887EA 265 EUR
613-1887
Robotic tips for BioTek™ pipetting systems
Pipette Tips
These automation tips for BioTek™ pipetting systems undergo a rigorous 15-point quality control program to ensure consistency thus providing accurate and precise performance achieved by superior tip straightness and a low coefficient of variation.
  • Sterile pipette tips are certified free of RNase, DNase, DNA, endotoxin, bioburden and pyrogen
  • Lot-tested during production on actual workstation for which they are created
  • Low percentage coefficient of variation (%CV)