Single-channel pipettes, mechanical, fixed / variable volume, Finnpipette™ F2
Supplier: Thermo Fisher Scientific
4642030 4642040 4642050 4642060 4642070 4642080 4642090 4642100 4642110 4652000 4652010 4652020 4652030 4652040 4652050 4652060 4642010 4642020 4652070 4652080 4652090 4652100 4652110 4652120 4642120 4642130 4652130 4652140
613-2698EA 483 EUR
613-2698 613-2699 613-2700 613-2701 613-2702 613-2703 613-2704 613-2705 613-2706 613-2707 613-2708 613-2709 613-2710 613-2711 613-2712 613-2713 613-2696 613-2697 613-2714 613-2715 613-2716 613-2717 613-2718 613-2719 613-1486 613-1487 613-1480 613-1481
Single-channel pipettes, mechanical, fixed / variable volume, Finnpipette™ F2
Pipettes Micropipettes
Finnpipette™ F2 offers superior robustness and durability even in the most demanding applications.
  • Fully autoclavable - No need to dismantle before autoclaving
  • Made of high-quality PVDF that withstands exposure to chemicals and UV light
  • Advanced Volume Gearing mechanism (AVG) - self-supporing modular mechanism increases accuracy, precision and durability, with light force and smaller increments for volume adjustment
  • Large, contrasted Ergovisio display with white numbers on black background for increased volume visibility and security
  • Soft-touch tip
More Product Information