Multi-channel pipettes, mechanical, variable volume, Proline® Plus
Supplier: Sartorius
728120 728130 728140 728220 728230 728240
613-5180EA 945 EUR
613-5180 613-5181 613-5182 613-5183 613-5184 613-5185
Multi-channel pipettes, mechanical, variable volume, Proline® Plus
Pipettes Micropipettes
The Proline® Plus mechanical pipettes are designed to offer comfort and quality for your everyday manual pipetting. They combine durable construction with ease and lightness of use, and are therefore a perfect choice for liquid-handling professionals and students.
  • Ergonomic design with light pipetting forces, comfortable handle and finger support for reduced risk of strain injuries
  • Optiload tip loading mechanism enables perfect tip sealing onto every individual tip cone
  • Minimised risk of contamination with Safe-Cone Filters and full autoclavability
  • Highly
More Product Information