VWR®, Pipette Tips for Rainin® LTS® Pipettes, Low Retention
Supplier: VWR Collection
613-5784EA 62 EUR
613-5784 613-5789
VWR®, Pipette Tips for Rainin® LTS® Pipettes, Low Retention
Pipette Tips
These tips are made with a special resin that reduces the potential for sample retention inside the tip. They help users reduce sample loss when performing protein research. These tips fit Rainin® LTS® pipettes.
  • Fit and eject with significantly less force than most conventional pipette tips
  • Ultra-fine points and super-clear resin for excellent viewing of sample
  • Free of RNase, DNase, endotoxins and trace metals
  • Tested for ATP
  • Autoclavable
Unlike pre-sterilised pipette tips from other brands, these tips use a validated sterile process (ISO) and can beMore Product Information