Single channel pipettes, mechanical, variable volume, PIPETMAN® L
Supplier: GILSON
FA10008 FA10007 FA10006M FA10003P FA10005P FA10006P FA10003M FA10005M FA10004P FA10004M FA10001M FA10001P FA10002M FA10002P F167350 F167550 F167370
613-5813EA 342 EUR
613-5813 613-5812 613-5805 613-5808 613-5810 613-5811 613-5802 613-5804 613-5809 613-5803 613-5800 613-5806 613-5801 613-5807 613-5890 613-5891 613-5892
Single channel pipettes, mechanical, variable volume, PIPETMAN® L
Pipettes Micropipettes
Designed for performance and comfort, PIPETMAN® L comes in a comprehensive range of models, adapted to your needs in the lab. PIPETMAN L air-displacement pipettes cover a full volume range. Comfortable, precise, accurate, and reliable, use PIPETMAN L pipettes confidently in your applications.
  • Convenient use
  • Secure pipetting
  • GLP compliance
Lightweight and balanced, PIPETMAN L combines an ergonomic design with minimized pipetting forces, giving you the ability to pipette longer without fatigue. Right-handed or left-handed? No problem. Adjust theMore Product Information