Pipetting system for 96- and 384-well plates, PLATEMASTER®
Supplier: GILSON
F110761 F110762
631-1375EA 16594 EUR
631-1375 613-5913
Pipetting system for 96- and 384-well plates, PLATEMASTER®
Automated Liquid Handling Systems
This compact, ergonomic system is an easy to use and accurate solution for high throughput pipetting of 96- and 384-well plates.
  • Helps to save valuable time, money and resources in the lab
  • Filling a 96-well plate requires less than 10 seconds
  • Fill a 384-well plate in less than a minute, in only 4 pipetting steps
  • Greatly reduces the number of pipetting steps necessary to fill a microplate, when compared to using regular multi-channel pipettes
  • Doesn't use a motor or require electricity, so it performs high-throughput pipetting silently
  • Reduce risk of human
More Product Information