Capillary pistons for Microman® pipettes
Supplier: GILSON
F148112 F148412 F148414 F148314 F148114 F148560 F148312 F148313 F148180 F148415 F148413 F148012 F148013 F148014 F148315 F148714 F148712 F148713
612-2545EA 0 EUR
612-2545 612-2549 612-2551 613-5936 612-7821 613-5940 612-7822 613-5935 613-5934 613-5939 613-5938 613-5930 613-5931 613-5932 613-5937 613-5943 613-5941 613-5942
Capillary pistons for Microman® pipettes
Pipette Tips
Designed to fit the Gilson MICROMAN® positive displacement pipette, Capillary Piston Tips are ideal for difficult-to-pipette samples including; viscous, volatile, dense, hot or cold, or hazardous liquids. The tips work like a syringe providing protection against pipette, sample, and user contamination, and produce accurate, reproducible, and precise results. Made of pure polypropylene, their slim fit allows you to use even the narrowest and deepest test tubes.
  • Accurate and precise pipetting
  • Protection against contamination
  • Convenient
Using CapillaryMore Product Information