Bottle top dispenser, Calibrex™ organo 525
Supplier: Socorex Isba
525.025 525.050 525.100 525.025FC 525.050FC 525.100FC 525.001 525.001FC 525.002.5 525.002.5FC 525.005 525.005FC 525.010 525.010FC
613-5976EA 480 EUR
613-5976 613-5978 613-5980 613-5977 613-5979 613-5981 613-5968 613-5969 613-5970 613-5971 613-5972 613-5973 613-5974 613-5975
Bottle top dispenser, Calibrex™ organo 525
Bottle Top Dispensers
Robust dosing instruments with excellent chemical resistance, high performance and simplified maintenance for safe and reproducible distribution ranging from 0,1 to 100 ml.
  • Ground glass or ceramic plunger for organic and non-crystallizing acid and base solutions
  • QR coded chemical compatibility
  • Integrated calibration key
  • Quick disassembling, no tool required
  • Autoclavable at 121 °C fully assembled
Flow control stopcok available, for priming without reagent loss or contamination, with locking position for safe dispenser transportation. Screw-lockingMore Product Information