Single-channel pipettes, mechanical, fixed / variable volume, Transferpette® S
Supplier: Brand
705891 705808 705816 705820 705862 705854 705864 705844 705838 705828 705868 705884 705882 705880 705878 705874 705873 705872 705870 705869 705890 705897 705896 705893 705892 DAKKS705844 DAKKS705862 DAKKS705874 DAKKS705880 DAKKS705882 DAKKS705873 DAKKS705870 DAKKS705878
613-6735EA 995 EUR
613-6735 613-6832 613-6833 613-6834 613-6835 613-6836 613-6837 613-6838 613-6839 613-6840 613-6841 613-6842 613-6843 613-6844 613-6845 613-6846 613-6847 613-6848 613-6849 613-6850 613-6734 613-6739 613-6738 613-6737 613-6736 BRNDDAKKS705844 BRNDDAKKS705862 BRNDDAKKS705874 BRNDDAKKS705880 BRNDDAKKS705882 BRNDDAKKS705873 BRNDDAKKS705870 BRNDDAKKS705878
Single-channel pipettes, mechanical, fixed / variable volume, Transferpette® S
Pipettes Micropipettes
The Transferpette® S air displacement pipette allows you to work efficiently and ergonomically with both small and large volumes. Due to its perfect ergonomics, the Transferpette® S provides a comfortable grip in any hand position, whether you are right- or left-handed, or have big or small hands.
  • True one-hand operation for right- and left-handed users: Easily set the volume, pipette and eject the tip
  • Colour code on volume adjustment wheel, shaft and volume-change protection
  • Low operating forces and a short stroke ensure relaxed pipetting, even during
More Product Information