Racked Pipette Tips for Universal Fit Pipettes
Supplier: Polycarbin
PC01-04-01 PC01-04-04 PC01-05-03 PC01-05-04 PC01-06-03 PC01-06-04 PC01-07-03 PC01-07-04
732-3980EA 775 EUR
732-3980 613-7223 732-3981 613-7224 732-3982 613-7225 732-3983 613-7226
Racked Pipette Tips for Universal Fit Pipettes
Pipette Tips
Polycarbin's pipette tip rack is made of 100% circular economy, post-consumer recycled content, combining high performance with a significantly reduced carbon footprint. Pipette tips do not contain recycled content, and are universal fit.
  • Manufactured in an ISO 9001 compliant facility
  • Carries a smaller carbon footprint
  • DNase, RNase and Pyrogen free
  • Sterile
Polycarbin's line of low-carbon lab products are designed for the sustainability-minded scientist. When designing our line of pipette tips, we focused on the tip box, or 'rack,' which is the mostMore Product Information