Axygen® HybridRack Pipette Tips
Supplier: Corning
ERF-10XT-R-S ERF-200NX-L-RS ERF-200NX-R-S ERF-20-L-R-S ERF-20-R-S ER-1000XT-R-S ER-10XT-R-S ER-200-R-S ER-300NX-R-S ER-300XT-R-S ERF-1000XT-L-R ERF-1000XT-R-S ERF-100-L-R-S ERF-100-R-S ERF-10XT-L-R-S ERF-300XT-L-RS ERF-300XT-R-S
613-7395EA 143 EUR
613-7395 613-7407 613-7398 613-7393 613-7396 613-7405 613-7401 613-7402 613-7403 613-7404 613-7406 613-7400 613-7394 613-7397 613-7392 613-7408 613-7399
Axygen® HybridRack Pipette Tips
Pipette Tips
Filtered, maxymum recovery, and sterile pipette tips in a truly sustainable packaging format that reduces plastic by 70% per case.
  • Patent pending design uses 70% less plastic than traditional tip rack, equal to six pounds of plastic saved per case
  • Paperboard base is easy to recycle with most local recycling programs
  • Made in the USA in an EngergyStar, ISO 14001, and ISO 13485 certified facility
  • Combination of the strong plastic deck and sturdy paperboard base allow for pipetting directly from the HybridRack, even with a mulit-channel instrument
  • Colour coded
More Product Information