Precision butyrometer, without stopper
Supplier: GERBER FUNKE
3150 3150-O 3150-C 3150-C-O
614-0088EA 17.5 EUR
614-0088 GERB3150-O GERB3150-C GERB3150-C-O
Precision butyrometer, without stopper
Butyrometers
For drinking vat milk, frosted rear scale wall, error tolerance 0,025%.