Pipette tips
Supplier: BIOTIX
2040 2042 2160 2107.Y 2102 2102.Y 2110.B 2100
614-1405EA 359 EUR
614-1405 614-1406 614-1413 BITX2107.Y BITX2102 BITX2102.Y 614-0087 BITX2100
Pipette tips
Pipette Tips
These pipette tips are made from virgin PP or exclusive S3 polymer.
  • Polymer tips retain virtually no fluids, allowing the user to dispense truly accurate sample volumes
  • Environmentally friendly packaging: some tips available in ESP reload systems
  • Tips are certified free of RNase, DNase, DNA and endotoxins
  • Graduated sides
The exclusive S3 polymer delivers to within 0,1% of indicated volume, eliminating sample hold-up and ensuring a more precise application. This proprietary technology is essential for quantitative assays where sample retention canMore Product Information