Lauric acid, Supelco®

Supplier: Merck
Warning

Synonyms: Dodecanoic acid, Dodecylic acid

MQ100

61609-5G
61609-5GEA 137 EUR
61609-5G
Lauric acid, Supelco®
Lauric acid
Formula: C₁₂H₂₄O₂
MW: 200.32 g/mol
Boiling Pt: 296
Melting Pt: 42…45 °C
Density: 0.875 g/cm³ (20 °C)
Flash Pt: >150 °C
Storage Temperature: Ambient
MDL Number: MFCD00002736
CAS Number: 143-07-7
EINECS: 205-582-1
Merck Index: 13,05400
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b61fb645a04ea22da93a7db1cfb44cf925f7e283&vid=0fcb82eedbbd50f1141aacef9994673dcd449a38&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR