Temperature and humidity loggers, EBI 20
Supplier: EBRO
1601-0010 1601-0011 1601-0042 1601-0043 1601-0046 1601-0047
620-1555EA 226 EUR
620-1555 620-1926 620-1748 620-1749 620-1751 620-1752
Temperature and humidity loggers, EBI 20
Data Loggers
These temperature and humidity loggers record data automatically. The LCD shows the measured value, battery level and unit of measure. These waterproof loggers with NTC probe provide reliable recordings during transport and storage.
  • Measurement frequency between 1 minute and 24 hours
  • One button for 'Start' and minimum/maximum values
  • Visual LED alarm
  • Memory mode: Endless measurement, start immediately until end of memory, start/stop measurement, start with key press
  • IP 67 protection
  • Applications: Transport, storage and process monitoring