VWR® Traceable®, Fast Response Digital Thermometer
Supplier: VWR Collection
620-2049EA 350 EUR
620-2049
VWR® Traceable®, Fast Response Digital Thermometer
Thermometers Digital Thermometers
This dual-channel thermometer allows the monitoring of two separate probes. The dual-display simultaneously shows the temperature of both probes or the temperature of one probe and the difference between the two (T1-T2). This thermometer can be used with all K-type thermocouple probes.
  • Serial computer output allows connection to computer or data logger for capturing and storing results
  • Memory recalls maximum/minimum readings
  • 'HOLD' button freezes the current display
Two fast response (<1 second) probes with 1,2 metre cable.
Certifications: More Product Information