VWR® Traceable®, Remote Alarm Thermo-Hygrometer
Supplier: VWR Collection
620-2486EA 139 EUR
620-2486 620-2487
VWR® Traceable®, Remote Alarm Thermo-Hygrometer
Hygrometers Digital Thermo-Hygrometers
These thermo-hygrometers are ideal for routine measurements, monitoring 'round the clock', all quality control needs, and critical experimental requirements. Perfect for use in hoods, classrooms, stockrooms, environmental chambers and chemical storage areas.
  • Triple-display shows min., max. and current humidity/temperature readings
  • Min./max. temperature and humidity memories
  • High/low alarm feature may be set by user in 1% RH or 1° increments
  • F/°C switchable
  • Remote humidity/temperature sensor on a 2,1 metre cable
* Temperature
** Humidity
***TemperatureMore Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable