VWR® TD20, TD21, TD22 Precise, Universal, Waterproof Thermometers
Supplier: VWR Collection
620-2641EA 118 EUR
620-2641 620-2642 620-2643
VWR® TD20, TD21, TD22 Precise, Universal, Waterproof Thermometers
Thermometers Digital Thermometers
Waterproof recalibratable thermometers for high precision measurements in liquids and in air, or for measurement of core temperatures (with insertion sensor); sensor handle and cable temperature range of up to 250 °C (permanent use temperature of 230 °C); laboratories, quality assurance, service, food, etc.
  • Short response time
  • Can be recalibrated at 0 °C and scale
  • Backlit 3-line display/overhead display at the push of a button
  • Alarm function
  • Waterproof (IP 65/IP 67)
  • Durable, long battery life
  • High quality sensors: Complete with PT1000 handheld sensor (up
More Product Information