CO₂ display for monitoring indoor air quality (IAQ)
Supplier: ROTRONIC INSTRUMENTS
CO2-DISPLAY
620-2757EA 580 EUR
620-2757
CO₂ display for monitoring indoor air quality (IAQ)
Monitoring Systems CO2 and Temperature Monitoring Systems
The wall-mounted or benchtop CO₂ display evaluates the quality of air with a combined measurement of CO₂, humidity and temperature.
  • HYGROMER® IN-1 humidity sensor
  • Adjustable, visual CO₂ indicator - CO₂ indicator for quick recognition of air quality
  • Easy recalibrate and adjustment function for humidity and CO₂
  • Large, easy to read display (date and time)
  • USB access for data download
The instrument can be configured directly with buttons and recorded data can be exported to a USB stick for analysis with the software package SW21. A high concentration ofMore Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable