Adhesion slides, Menzel Gläser, SuperFrost® Plus Gold
Supplier: Epredia
K5800AMNZ72
630-1324EA 134 EUR
630-1324
Adhesion slides, Menzel Gläser, SuperFrost® Plus Gold
Slides Adhesive Slides
Glass adhesive technology, which first attracts, and then chemically bonds fresh or formalin-fixed frozen tissue sections firmly to the surface.
  • Ground edges (90°) and frosted end for labelling
  • Attaches difficult to bond tissue sections
  • Washed and polished
Suitable for special stains, immunocytochemicals and in situ DNA hybridisation techniques.