Stereo zoom microscopes, SMZ-171
Supplier: MOTIC
1100200600766 1100200600751 1100200600754 1100200600758 1100200600761 1100200600762
630-1807EA 1700 EUR
630-1807 630-2024 630-2025 630-2026 630-2027 630-2028
Stereo zoom microscopes, SMZ-171
Microscopes
The SMZ-171 modular system enables you to create your own microscope by choosing a head, a holder and a stand.
  • Greenough optical system
  • Working distance of 110 mm
  • Transmitted illumination with tilting reflector
The optical performance (max. 460 lp/mm), combined with an expanded accessory line, make the SMZ-171 the instruments for demanding requirements, both for routine and research fields. They deliver clear, distortion-free, 3-D images for biological and medical preparations, industrial quality control and inspection.