LED illumination units for fluorescence, CoolLED pE-200 and pE-300
Supplier: COOLLED
PE300WDSB71E30 PE300WDSB20E30 PE300WDSB30E30 PE300WDSB40E30 PE300WDSB50E30 PE300WDSB22E30 PE300WDSB72E30
630-2494EA 0 EUR
630-2494 630-2489 630-2491 630-2492 630-2493 630-2490 CLEDPE300WDSB72E30
LED illumination units for fluorescence, CoolLED pE-200 and pE-300
Lights LED Lights, Lamps and Bulbs
These LED illuminators are designed to replace high pressure mercury lamps used on fluorescence microscopes. They can be specified with two or three LED wavelength combinations, respectively. No mercury, instant on/off, no bulb changing, no downtime, no warm up, no unwanted UV and reduced photobleaching. You can also benefit from better contrast for sharper images and seeing deeper into cells. They are suitable for routine screening, tissue culture, diagnostics, confocal and clinical.
  • Intensity control: 0 to 100% in 1% increments (independent for each LED
More Product Information