LED Illumination Systems, EasyLED
Supplier: SCHOTT AG Lighting and imaging
600101 600121 600121-SETC 600121-SETSA
630-2662EA 445 EUR
630-2662 630-2663 630-3491 630-3492
LED Illumination Systems, EasyLED
Lights LED Lights, Lamps and Bulbs
The SCHOTT EasyLED portfolio for stereo microscopy delivers precise LED light for a wide variety of applications, such as laboratory and diagnostics, industrial applications, education, or forensic science. The series offers robust design and easy handling in a compact device, with a range of accessories for specific requirements.
  • Powerful luminance; Intense light output for specialised tasks
  • Maintenance-free LED lifetime of 50000 hours
  • Color temperature: approximately 5600 K
  • Low power consumption; smart engineering leads to optimized energy use
  • Compatible
More Product Information